Signs, picto-pack
Supplier: Brady
223863
BRDY223863EA 0 SGD
BRDY223863
Signs, picto-pack
Signs
Picto-Pack is a sensible combination of a wide range of signs and labels made from various materials.
  • PE: Self-adhesive, laminated polyester
  • PP: Polypropylene (not self-adhesive)
  • V: Vinyl (photoluminescent edge)
  • AL: Aluminium (photoluminescent edge)