Economy spill kits in duffel bag, PIG® Haz-Mat
Supplier: NEW PIG
KIT323 KIT342
NEWPKIT323EA 1010 SGD
NEWPKIT323 NEWPKIT342
Economy spill kits in duffel bag, PIG® Haz-Mat
Spill Control Materials
This kit includes four chemical spill packs in one large, sturdy sports bag. Just open kit, use the absorbents, then throw them in the temporary disposal bags included.
  • Makes a great vehicle spill kit
  • Great for multiple uses or locations
  • PIG® absorbents are proven for rapid response
  • Bold spill Kit lettering for quick identification
  • PIG® HazMat socks stop spreading spills; PIG® HazMat pads absorb quickly
  • Temporary disposal bags help make cleanup easier
PIG® HazMat absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulphuric acid and 30% sodium hydroxide. Keep a refill handy to instantly replace used contents.
Delivery information: Kit includes 4×refill for PIG® HazMat economy spill kits in duffel bag, 3×PIG® HazMat chemical absorbent sock (76×1168 mm), 48×PIG® HazMat mat pad (381×508 mm), 8×polyethylene disposal bags (457×762) and instructions. Refill includes 3×PIG® HazMat chemical absorbent sock (76×1168 mm), 12×PIG® HazMat mat pad (381×508 mm), 2×polyethylene disposal bags (457×762) and instructions.