Crocetin free acid (from Gardenia jasminoides ellis)

Supplier: MP Biomedicals

Synonyms: 2,6,11,15-Tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedoic acid, (2E,4E,6E,8E,10E,12E,14E)-2,6,11,15-Tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid, 8,8´-Diapocarotenedioic acid

0219354305 0219354325
ICNA0219354305EA 149 SGD
ICNA0219354305 ICNA0219354325
Crocetin free acid (from Gardenia jasminoides ellis)
Crocetin

Crocetin inhibitor is a natural carotenoid dicarboxylic acid that is found in the crocus flower and Gardenia jasminoides.


Crocetin was found to inhibit cell cycle progression at the G2-M phase and pancreatic tumor formation in nude mice. It has been shown to improve postshock recovery of cellular adenosine triphosphate (ATP). It also inhibits pancreatic cancer cell proliferation and tumor progression in a xenograft mouse mode.


Crocetin may provide neuroprotection in rats by reducing the production of various neurotoxic molecules.

Formula: C₂₀H₂₄O₄
MW: 328,41 g/mol
Melting Pt: 258…259 °C
MDL Number: MFCD00017359
CAS Number: 27876-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR