Bottles, wide neck, clear, french square
Supplier: BERLIN PACKAGING, LLC
GLC-01311 GLC-01363 GLC-01387
BERLGLC-01311EA 180 SGD
BERLGLC-01311 BERLGLC-01363 BERLGLC-01387
Bottles, wide neck, clear, french square
Bottles Glass Bottles
Made of glass, clear.
  • Square shape saves on shelf and storage space
  • Graduated in both ml and oz.
  • Provide maximum content visibility
These clear glass French Square bottles with wide mouth opening provide maximum content visibility and is ideal for mixing, storing and sampling.