VWR®, Vaccum Ovens, digital
Supplier: VWR Collection
97027-674 97027-664
VWRU97027-674EA 35060 SGD
VWRU97027-674 VWRU97027-664
VWR®, Vaccum Ovens, digital
Ovens Vacuum Ovens
These vacuum ovens are built with a stainless steel chamber for exceptional durability.
  • Fully programmable Watlow temperature control
  • Capable of 40-step ramp and soak profiles or 4 files With 10 steps per file
  • Digital vacuum gauge
  • RS485 communication
The doors on these units have positive latch handles with spring-loaded glass to facilitate a good vacuum seal without hinge binds that shorten the gasket life. Versatile based on the selection of gaskets and a small benchtop footprint.
The unique cross-flow ventilation forces inert gas to fill theMore Product Information