Brefeldin A 98, powder_white Molecular_Biology

Supplier: MP Biomedicals
Danger

Synonyms: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural

0219480205 0219480210
ICNA0219480205EA 597 SGD
ICNA0219480205 ICNA0219480210
Brefeldin A 98, powder_white Molecular_Biology
Brefeldin A

Storage: Store at -20 °C.
Brefeldin A is a fungal metabolite which is a macrocyclic lactone exhibiting a wide range of antibiotic activity. Produced by Penicillium brefeldianum. Blocks binding of the cytosolic coat protein b-COP and ARF to Golgi membranes mediated by protein G. Also blocks protein transportation into post-Golgi compartments. It activates the sphingomyelin cycle. Brefeldin A mediated apoptosis has been observed in human tumor cells.
Brefeldin A reversibly inhibits the intracellular translocation of proteins in eukaryotes, e.g., during transport of proteins to the cell surface for secretion or expression. It has been reported to block the response of cultured cells to cholera toxin. In HepG2 cells, BFA induces two blocks in the secretory pathway; one at the level of the endoplasmic reticulum-Golgi juncture and the other in the trans-Golgi network. Brefeldin A is used in the studies of Brefeldin A-inhibited Guanine Nucleotide-exchange Protein, BIG2, Regulates the Constitutive Release of TNFR1 Exosome-like Vesicles.
Brefeldin A (BFA) is a fungal metabolite which disrupts the structure and function of the Golgi apparatus. BFA is an activator of the sphingomyelin cycle. Brefeldin A-mediated apoptosis has been observed in human tumor cells.


  • Molecular Biology Reagent Grade
  • Presentation: White Powder
  • Soluble in chloride, methylene, ethanol (5 mg/mL-clear, colorless solution), methanol (5 mg/mL - clear, colorles solution), or DMSO (20 mg/mL).

Formula: C₁₆H₂₄O₄
MW: 280.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00083258
CAS Number: 20350-15-6
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR