Blocks for digital dry block incubators
Supplier: BOEKEL SCIENTIFIC
110010 110013 110016 110020 110025 110035 110040 110045 110051 110096 110099
BOEI110010EA 336 SGD
BOEI110010 BOEI110013 BOEI110016 BOEI110020 BOEI110025 BOEI110035 BOEI110040 BOEI110045 BOEI110051 BOEI110096 BOEI110099
Blocks for digital dry block incubators
Block Heaters Blocks
Interchangeable heating dry block modules are made of solid aluminium with a black anodised finish. These are features with alpha/numerical labelled rows and columns for easy vessel identification.