Erbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Scientific
Danger


Packaged under argon.

12388.FF 12388.FI
ALFA12388.FFEA 0 SGD
ALFA12388.FF ALFA12388.FI
Erbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.025 mm (0.001 in)
Erbium

Purity: 99.9% CAS No.: 7440-52-0 Physical Form: ≈0.14g/25x25mm, Packaged under argon UN No.: 3178 MDL No.: MFCD00010987

Formula: Er
MW: 167,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1
UN: 3178
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR