Europium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific
Danger

10175.FF 10175.FI
ALFA10175.FFEA 1850 SGD
ALFA10175.FF ALFA10175.FI
Europium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Europium

Purity: 99.9% CAS No.: 7440-53-1 Physical Form: ≈0.82g/25x25mm, Packaged in mylar under argon Appearance: Grey Odour: Odourless UN No.: 3208 MDL No.: MFCD00010992

Formula: Eu
MW: 151,96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010992
CAS Number: 7440-53-1
EINECS: 231-161-7
UN: 3208
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR