Samarium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.75 mm (0.03 in)

Supplier: Thermo Scientific

43218.FF 43218.FI
ALFA43218.FFEA 584 SGD
ALFA43218.FF ALFA43218.FI
Samarium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.75 mm (0.03 in)
Samarium

Purity: 99.9% CAS No.: 7440-19-9 Physical Form: ≈3.7g/25x25mm, Packaged in mylar under argon Odour: Odourless MDL No.: MFCD00011233

Formula: Sm
MW: 150,36 g/mol
Boiling Pt: 1900 °C (1013 hPa)
Melting Pt: 1076,85 °C
Density: 7,42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
Merck Index: 12,08498

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR