Gold ≥99.96% (metals basis), flakes APS 1,5-3,0 µm
Supplier: Thermo Scientific
42151.03
42151.06
ALFA42151.03EA
0
SGD
ALFA42151.03
ALFA42151.06
Gold ≥99.96% (metals basis), flakes APS 1,5-3,0 µm
Gold
Purity: 99.96%
CAS No.: 7440-57-5
MDL No.: MFCD00003436
Formula:
Au Boiling Pt: 2800 °C (1013 hPa) Melting Pt: 1063 °C Density: 19,3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...