Tantalum ≥99.95% (metals basis excluding Nb), foil, annealed, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Scientific
10355.FF
10355.FI
10355.GH
10355.GW
ALFA10355.FFEA
0
SGD
ALFA10355.FF
ALFA10355.FI
ALFA10355.GH
ALFA10355.GW
Tantalum ≥99.95% (metals basis excluding Nb), foil, annealed, Thickness: 0.5 mm (0.02 in)
Tantalum
Purity: 99.95%
CAS No.: 7440-25-7
Physical Form: ≈5.19g/25x25mm
Odour: Odourless
MDL No.: MFCD00011252
Formula:
Ta Boiling Pt: 5427 °C (1013 hPa) Melting Pt: 2996 °C Density: 16,69 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...