4'-Hydroxychalcone ≥97%
Supplier: Thermo Scientific
|
Warning
|
A10838.06
A10838.14
ALFAA10838.06EA
97.9
SGD
ALFAA10838.06
ALFAA10838.14
4'-Hydroxychalcone ≥97%
4'-Hydroxychalcone
Purity: 97%
CAS No.: 2657-25-2
Molecular Formula: C15H12O2
Formula Weight: 224.26
Melting Point: 175-177°
Beilstein Reference: 2049150
Formula:
C₁₅H₁₂O₂ Melting Pt: 175…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016484 CAS Number: 2657-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...