(±)-1-Octyn-3-ol ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Oct-1-yn-3-ol
A13267.09
A13267.18
A13267.30
ALFAA13267.09EA
0
SGD
ALFAA13267.09
ALFAA13267.18
ALFAA13267.30
(±)-1-Octyn-3-ol ≥98%
(±)-1-Octyn-3-ol
Purity: 98%
CAS No.: 818-72-4
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 63°(145°F)
Refractive Index: 1.4420
Melting Point: -60°
Boiling Point: 180-181°
Beilstein Reference: 1098642
MDL No.: MFCD00004588
Formula:
C₈H₁₄O MW: 126,2 g/mol Boiling Pt: 180…181 °C Melting Pt: –60 °C Density: 0,865 Flash Pt: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004588 CAS Number: 818-72-4 EINECS: 212-455-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...