(±)-4-Heptyn-2-ol ≥97%
Supplier: Thermo Scientific
Warning
|
L11292.06
L11292.14
ALFAL11292.06EA
139
SGD
ALFAL11292.06
ALFAL11292.14
(±)-4-Heptyn-2-ol ≥97%
(±)-4-Heptyn-2-ol
Purity: 97%
CAS No.: 19781-81-8
Molecular Formula: C7H12O
Formula Weight: 112.17
Flash Point: 62°(143°F)
Refractive Index: 1.4515
Boiling Point: 67-68°/14mm
Beilstein Reference: 2232633
MDL No.: MFCD00041588
Formula:
C₇H₁₂O MW: 112,17 g/mol Boiling Pt: 67…68 °C (14 mmHg) Density: 0,880 Flash Pt: 62 °C (143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041588 CAS Number: 19781-81-8 EINECS: 243-309-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...