Cobalt(II) fluoride tetrahydrate ≥99.99% (metals basis), Puratronic®

Supplier: Thermo Scientific
Danger

Synonyms: Cobaltous fluoride tetrahydrate

10693.06 10693.14
ALFA10693.06EA 156 SGD
ALFA10693.06 ALFA10693.14
Cobalt(II) fluoride tetrahydrate ≥99.99% (metals basis), Puratronic®
Cobalt (II) fluoride tetrahydrate

Purity: 99.99% CAS No.: 13817-37-3 Molecular Formula: CoF2.4H2O Formula Weight: 168.99 (96.93anhy) Physical Form: Powder Density: 2.19 Appearance: Pink Odour: Odourless Melting Point: 200° dec. Merck Reference: 14,2440 UN No.: 2923 MDL No.: MFCD00149656

Formula: CoF₂·4H₂O
MW: 168,99 g/mol
Melting Pt: 200 °C (dec.)
Density: 2,19
Storage Temperature: Ambient
MDL Number: MFCD00149656
CAS Number: 13817-37-3
EINECS: 233-061-9
UN: 2923
ADR: 8,III
Merck Index: 14,2440

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR