(S)-(+)-2-Heptanol ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(S)-(+)-Pentyl methyl carbinol, (S)-Heptan-2-ol, (S)-(+)-Methyl pentyl carbinol, (S)-(+)-2-Hydroxy heptane
33795.03
33795.MD
33795.06
ALFA33795.03EA
0
SGD
ALFA33795.03
ALFA33795.MD
ALFA33795.06
(S)-(+)-2-Heptanol ≥98%
(S)-(+)-2-Heptanol
Purity: 98%
CAS No.: 6033-23-4
Molecular Formula: C7H16O
Formula Weight: 116.20
Flash Point: 59°(138°F)
Physical Form: Liquid
Density: 0.815
Refractive Index: 1.4210
Boiling Point: 160-162°
Merck Reference: 14,4662
UN No.: 1987
MDL No.: MFCD00065003
Formula:
C₇H₁₆O MW: 116,2 g/mol Boiling Pt: 160…162 °C Density: 0,815 Flash Pt: 59 °C (138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00065003 CAS Number: 6033-23-4 UN: 1987 ADR: 3,III Merck Index: 14,4662 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...