(R)-(+)-1-Octyn-3-ol ≥98%
Supplier: Thermo Scientific
|
Warning
|
L19045.03
L19045.MD
ALFAL19045.03EA
423
SGD
ALFAL19045.03
ALFAL19045.MD
(R)-(+)-1-Octyn-3-ol ≥98%
(R)-(+)-1-Octyn-3-ol
Purity: 98+%
CAS No.: 32556-70-0
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 63°(145°F)
Refractive Index: 1.4420
Boiling Point: 100°/20mm
Beilstein Reference: 2037704
Optical Rotation: +9.9° (neat)
MDL No.: MFCD00192370
Formula:
C₈H₁₄O MW: 126,2 g/mol Boiling Pt: 100 °C (20 mmHg) Density: 0,866 Flash Pt: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00192370 CAS Number: 32556-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...