Butyramide ≥98%

Supplier: Thermo Scientific
Warning

A12494.18 A12494.22 A12494.36
ALFAA12494.18EA 84.9 SGD
ALFAA12494.18 ALFAA12494.22 ALFAA12494.36
Butyramide ≥98%
Butyramide

Purity: 98% CAS No.: 541-35-5 Molecular Formula: CH3CH2CH2CONH2 Formula Weight: 87.12 Melting Point: 113-116° Boiling Point: 216° Merck Reference: 14,1592 MDL No.: MFCD00041894

Formula: C₄H₉NO
MW: 87,12 g/mol
Boiling Pt: 216 °C
Melting Pt: 113…116 °C
Density: 1,032
Storage Temperature: Ambient
MDL Number: MFCD00041894
CAS Number: 541-35-5
EINECS: 208-776-4
Merck Index: 14,1592

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR