1-Naphthyl butyrate ≥98%
Supplier: Thermo Scientific
Warning
|
Synonyms:
Butyric acid 1-naphthyl ester, α-Naphthyl butyrate, Butanoic acid α-naphthyl ester, Butanoic acid 1-naphthyl ester, Butyric acid α-naphthyl ester
Purity: 98+%
CAS No.: 3121-70-8
Molecular Formula: C14H14O2
Formula Weight: 214.27
Flash Point: 62°(143°F)
Refractive Index: 1.5710
Boiling Point: 125-130°/1mm
Beilstein Reference: 2558646
MDL No.: MFCD00003923
Formula:
C₁₄H₁₄O₂ MW: 214,26 g/mol Boiling Pt: 125…130 °C (1 mmHg) Melting Pt: 95,5…96.5 °C Density: 1,090 Flash Pt: 62 °C (143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003923 CAS Number: 3121-70-8 EINECS: 221-500-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...