Cesium acetate ≥99.9% (metals basis)
Supplier: Thermo Scientific
Synonyms:
Acetic acid cesium salt
12929.14
12929.22
ALFA12929.14EA
130
SGD
ALFA12929.14
ALFA12929.22
Cesium acetate ≥99.9% (metals basis)
Cesium acetate
Purity: 99.9%
CAS No.: 3396-11-0
Molecular Formula: CsOOCCH3
Formula Weight: 191.95
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00013056
Formula:
H₃CCOOCs MW: 191,95 g/mol Melting Pt: 194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013056 CAS Number: 3396-11-0 EINECS: 222-248-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...