Rubidium acetate ≥99.8% (metals basis)

Supplier: Thermo Scientific

Synonyms: Acetic acid rubidium salt

12890.09 12890.18
ALFA12890.09EA 119 SGD
ALFA12890.09 ALFA12890.18
Rubidium acetate ≥99.8% (metals basis)
Rubidium acetate

Purity: 99.8% CAS No.: 563-67-7 Molecular Formula: RbOOCCH3 Formula Weight: 144.52 Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00013058

Formula: H₃CCOORb
MW: 144,51 g/mol
Melting Pt: 246 °C
Storage Temperature: Ambient
MDL Number: MFCD00013058
CAS Number: 563-67-7
EINECS: 209-255-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR