1-Iodo-4,4,4-trifluorobutane ≥98% stabilised

Supplier: Thermo Scientific
Warning

Synonyms: 4-Iodo-1,1,1-trifluorobutane, 1,1,1-Trifluoro-4-iodobutane

Stabilization: Stabilised with copper

B23811.03 B23811.06
ALFAB23811.03EA 0 SGD
ALFAB23811.03 ALFAB23811.06
1-Iodo-4,4,4-trifluorobutane ≥98% stabilised
1-Iodo-4,4,4-trifluorobutane

Stabilized with copper


Purity: 98+% CAS No.: 461-17-6 Molecular Formula: C4H6F3I Formula Weight: 237.99 Flash Point: 100°(212°F) Refractive Index: 1.4335 Boiling Point: 126-127° Beilstein Reference: 1736064 MDL No.: MFCD00041543

Formula: C₄H₆F₃I
MW: 237,99 g/mol
Boiling Pt: 126…127 °C
Density: 1,85
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041543
CAS Number: 461-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR