4,4,4-Trifluoro-1-butanol ≥97%
Supplier: Thermo Scientific
|
Warning
|
B21460.03
B21460.06
ALFAB21460.03EA
0
SGD
ALFAB21460.03
ALFAB21460.06
4,4,4-Trifluoro-1-butanol ≥97%
4,4,4-Trifluoro-1-butanol
Purity: 97%
CAS No.: 461-18-7
Molecular Formula: CF3(CH2)3OH
Formula Weight: 128.09
Flash Point: 52°(125°F)
Refractive Index: 1.3430
Boiling Point: 122-124°
Beilstein Reference: 1736068
UN No.: 1993
MDL No.: MFCD00041483
Formula:
C₄H₇F₃O MW: 128,09 g/mol Boiling Pt: 122…124 °C Density: 1,193 Flash Pt: 52 °C (125 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041483 CAS Number: 461-18-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...