N,N-Dimethyldodecylamine ≥90%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
N,n-dimethyldodecylamine, Dodecyldimethylamine
B24145.0B
B24145.18
ALFAB24145.0BEA
1180
SGD
ALFAB24145.0B
ALFAB24145.18
N,N-Dimethyldodecylamine ≥90%
N,N-Dimethyldodecylamine
Purity: 90+%
CAS No.: 112-18-5
Molecular Formula: CH3(CH2)11N(CH3)2
Formula Weight: 213.41
Flash Point: 118°(244°F)
Refractive Index: 1.4375
Melting Point: -20°
Boiling Point: 110-112°/3mm
UN No.: 2735
MDL No.: MFCD00008970
Formula:
C₁₄H₃₁N MW: 213,41 g/mol Boiling Pt: 110…112 °C (3 mmHg) Melting Pt: –20 °C Density: 0,787 Flash Pt: 118 °C (244 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008970 CAS Number: 112-18-5 EINECS: 203-943-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...