1-Pentyl isothiocyanate 96%

Supplier: Thermo Scientific
Danger

A14972.14 A14972.06
ALFAA14972.14EA 0 SGD
ALFAA14972.14 ALFAA14972.06
1-Pentyl isothiocyanate 96%
1-Pentyl isothiocyanate

Purity: 96% CAS No.: 629-12-9 Molecular Formula: CH3(CH2)4NCS Formula Weight: 129.23 Refractive Index: 1.4965 Odour: Pungent Boiling Point: 191-193° Beilstein Reference: 1746691 UN No.: 2922 MDL No.: MFCD00014444

Formula: C₆H₁₁NS
MW: 129,23 g/mol
Boiling Pt: 191…193 °C
Density: 0,930
Storage Temperature: Ambient
MDL Number: MFCD00014444
CAS Number: 629-12-9
EINECS: 211-075-6
UN: 2922
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR