Membrane filters, cellulose acetate
Supplier: ADVANTEC
C045A047A C020A013A C020A025A C020A037A C020A047A C020A090C C020A142C C020A293C C020A330R C020G047A C045A013A C045A025A C045A037A C045A090C C045A102C C045A142C C045A293C C045A330R C045G047A C045G085A C080A013A C080A025A C080A037A C080A047A C080A090C C080A142C C080A293C C300A013A C300A025A C300A037A C300A047A C300A090C C300A142C C300A293C
ADVTC045A047AEA 0 SGD
ADVTC045A047A ADVTC020A013A ADVTC020A025A ADVTC020A037A ADVTC020A047A ADVTC020A090C ADVTC020A142C ADVTC020A293C ADVTC020A330R ADVTC020G047A ADVTC045A013A ADVTC045A025A ADVTC045A037A ADVTC045A090C ADVTC045A102C ADVTC045A142C ADVTC045A293C ADVTC045A330R ADVTC045G047A ADVTC045G085A ADVTC080A013A ADVTC080A025A ADVTC080A037A ADVTC080A047A ADVTC080A090C ADVTC080A142C ADVTC080A293C ADVTC300A013A ADVTC300A025A ADVTC300A037A ADVTC300A047A ADVTC300A090C ADVTC300A142C ADVTC300A293C
Membrane filters, cellulose acetate
Filters Membrane Filters
Composed of cellulose di- and triacetate, these filters exhibit low static charge and high strength.
  • Extremely low aqueous extractables (0,1 wt%)
  • Good resistance to heat and low molecular weight alcohols
Can be sterilised repeatedly without loss of integrity or change in bubble point. Use for filtration of enzyme solutions, diagnostic cytology, or receptor binding studies.
Provides better heat resistance up to 180 °C and may be repeatedly autoclaved without loss of integrity.