In-line filter holders, PFA
Supplier: ADVANTEC
43307010 43307020
ADVT43307010EA 470 SGD
ADVT43307010 ADVT43307020
In-line filter holders, PFA
Filter Holders
These filter holders are ideal for filtering small volumes of high purity acids, alkalis or organic solvents. They also sterilise and ultraclean liquids under positive pressure.
  • Superior chemical compatibility
  • Dual support screens allow for flow in either direction
  • With exterior locking ring
Locking ring ensures quick assembly without tearing the membrane filter or releasing lines. Body and support screens are made of PFA.
Note: PFA47 has an air/bleed vent.