(2-Chloro-6-fluorophenyl)acetic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Chloro-6-fluorophenylacetic acid

A10460.06 A10460.14 A10460.22
ALFAA10460.06EA 0 SGD
ALFAA10460.06 ALFAA10460.14 ALFAA10460.22
(2-Chloro-6-fluorophenyl)acetic acid ≥98%
(2-Chloro-6-fluorophenyl)acetic acid

Purity: 98% CAS No.: 37777-76-7 Molecular Formula: C8H6ClFO2 Formula Weight: 188.59 Melting Point: 120-124° Beilstein Reference: 4401142

Formula: (C₆H₃ClF)CH₂COOH
MW: 188,59 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 120…124 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004319
CAS Number: 37777-76-7
EINECS: 253-661-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR