pH indicator papers
Supplier: ADVANTEC
UNIV-R 07010080 07010070 07010040 07010140 07010030 07010060 07010100 07010010 07010050 07010110 07010090 07010130 07010150 07010020 UNIV-B 07011010 07011020 W-R
ADVTUNIV-REA 31.4 SGD
ADVTUNIV-R ADVT07010080 ADVT07010070 ADVT07010040 ADVT07010140 ADVT07010030 ADVT07010060 ADVT07010100 ADVT07010010 ADVT07010050 ADVT07010110 ADVT07010090 ADVT07010130 ADVT07010150 ADVT07010020 ADVTUNIV-B ADVT07011010 ADVT07011020 ADVTW-R
pH indicator papers
Test Papers pH Indicator Papers and Test Strips
Various types of indicator papers cover a wide pH range to allow for good accuracy of determining pH value of a solution.
High sensitivity and sharp color change. Narrow pH unit graduation: 0,2 – 0,4 (UNIV/ WR: 1,0 – 2,0 units).
Packaging: Booklet of 20 strips, 7×70 mm; Roll, 9 mm wide in dispenser.