Test papers, litmus
Supplier: ADVANTEC
07020020 07020010
ADVT07020020EA 22 SGD
ADVT07020020 ADVT07020010
Test papers, litmus
Test Papers Indicator Papers
General detection for whether solution is acidic (blue) or alkaline (red).
Color changes instantaneously, compare to chart for determination.
Packaging: Booklet of 20 strips, 7×70 mm.