Diethyl fumarate ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Fumaric acid diethyl ester

A16214.0E A16214.36
ALFAA16214.0EEA 0 SGD
ALFAA16214.0E ALFAA16214.36
Diethyl fumarate ≥98%
Diethyl fumarate

Purity: 98% CAS No.: 623-91-6 Molecular Formula: C8H12O4 Formula Weight: 172.18 Flash Point: 91°(195°F) Refractive Index: 1.4400 Melting Point: ca 0° Boiling Point: 98-100°/14mm Beilstein Reference: 775347 MDL No.: MFCD00064455

Formula: C₈H₁₂O₄
MW: 172,18 g/mol
Boiling Pt: 98…100 °C (14 mmHg)
Melting Pt: ∼ 0 °C
Density: 1,050
Flash Pt: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064455
CAS Number: 623-91-6
EINECS: 210-819-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR