cis-3-Hepten-1-ol ≥97%
Supplier: Thermo Scientific
Warning
|
A19172.06
A19172.14
ALFAA19172.06EA
119
SGD
ALFAA19172.06
ALFAA19172.14
cis-3-Hepten-1-ol ≥97%
cis-3-Hepten-1-ol
Purity: 97%
CAS No.: 1708-81-2
Molecular Formula: C7H14O
Formula Weight: 114.18
Flash Point: 68°(154°F)
Refractive Index: 1.444
Boiling Point: 92°/60mm
Beilstein Reference: 1719803
MDL No.: MFCD00274760
Formula:
C₇H₁₄O MW: 114,19 g/mol Boiling Pt: 92 °C (60 mmHg) Density: 0,85 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00274760 CAS Number: 1708-81-2 EINECS: 216-964-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...