trans-2-Bromocinnamic acid ≥98%

Supplier: Thermo Scientific
Warning

A11381.06 A11381.14
ALFAA11381.06EA 0 SGD
ALFAA11381.06 ALFAA11381.14
trans-2-Bromocinnamic acid ≥98%
trans-2-Bromocinnamic acid

Purity: 98+% CAS No.: 7345-79-1 Molecular Formula: C9H7BrO2 Formula Weight: 227.06 Odour: Odourless Melting Point: 216-219° Beilstein Reference: 2357933 MDL No.: MFCD00016836

Formula: C₉H₇BrO₂
MW: 227,06 g/mol
Melting Pt: 216…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00016836
CAS Number: 7499-56-1
EINECS: 231-353-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR