3-Bromo-4-fluorocinnamic acid ≥98%

Supplier: Thermo Scientific
Warning

A14526.06 A14526.14
ALFAA14526.06EA 0 SGD
ALFAA14526.06 ALFAA14526.14
3-Bromo-4-fluorocinnamic acid ≥98%
3-Bromo-4-fluorocinnamic acid

Purity: 98% CAS No.: 160434-49-1 Molecular Formula: C9H6BrFO2 Formula Weight: 245.05 Melting Point: 193-195° Beilstein Reference: 8977970

Formula: C₉H₆BrFO₂
MW: 245,05 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 193…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00143095
CAS Number: 160434-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR