4-Bromo-2-fluorocinnamic acid ≥98%

Supplier: Thermo Scientific
Warning

A16028.06 A16028.14
ALFAA16028.06EA 0 SGD
ALFAA16028.06 ALFAA16028.14
4-Bromo-2-fluorocinnamic acid ≥98%
4-Bromo-2-fluorocinnamic acid

Purity: 98% CAS No.: 149947-19-3 Molecular Formula: C9H6BrFO2 Formula Weight: 245.05 Melting Point: 216-221° MDL No.: MFCD00143267

Formula: C₉H₆BrFO₂
MW: 245,05 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 216…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00143267
CAS Number: 149947-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR