3,4-Dimethoxycinnamic acid predominantly trans 99%

Supplier: Thermo Scientific
Warning

Synonyms: 3,4-Dimethoxycinnamic Acid

A10356.14 A10356.22
ALFAA10356.14EA 0 SGD
ALFAA10356.14 ALFAA10356.22
3,4-Dimethoxycinnamic acid predominantly trans 99%
3,4-Dimethoxycinnamic acid

Purity: 99% CAS No.: 2316-26-9 Molecular Formula: C11H12O4 Formula Weight: 208.21 Melting Point: 179-183° Beilstein Reference: 1533435

Formula: C₁₁H₁₂O₄
MW: 208,21 g/mol
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00004387
CAS Number: 2316-26-9
EINECS: 219-025-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR