4-Methyl-1-pentyne-3-ol ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Hydroxy-4-methyl-1-pentyne, 4-Methylpent-1-yn-3-ol
L05318.09
L05318.18
ALFAL05318.09EA
0
SGD
ALFAL05318.09
ALFAL05318.18
4-Methyl-1-pentyne-3-ol ≥97%
4-Methyl-1-pentyne-3-ol
Purity: 97%
CAS No.: 565-68-4
Molecular Formula: C6H10O
Formula Weight: 98.15
Flash Point: 41°(105°F)
Refractive Index: 1.4360
Odour: Recognizable
Boiling Point: 133-134°
Beilstein Reference: 1698762
UN No.: 1987
MDL No.: MFCD00039847
Formula:
C₆H₁₀O MW: 98,14 g/mol Boiling Pt: 133…134 °C Density: 0,929 Flash Pt: 41 °C (105 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039847 CAS Number: 565-68-4 EINECS: 209-287-9 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...