9,10-Dichloroanthracene ≥97%
Supplier: Thermo Scientific
|
Warning
|
A12823.06
A12823.14
ALFAA12823.06EA
157
SGD
ALFAA12823.06
ALFAA12823.14
9,10-Dichloroanthracene ≥97%
9,10-Dichloroanthracene
Purity: 97%
CAS No.: 605-48-1
Molecular Formula: C14H8Cl2
Formula Weight: 247.12
Melting Point: 210-215°
Beilstein Reference: 1912108
Formula:
C₁₄H₈Cl₂ MW: 247,12 g/mol Boiling Pt: 366 °C (1013 hPa) Melting Pt: 210…215 °C Density: 1,525 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001246 CAS Number: 605-48-1 EINECS: 210-087-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...