3-Bromo-5-nitrobenzoic acid ≥99%

Supplier: Thermo Scientific
Warning

L19633.03 L19633.MD
ALFAL19633.03EA 0 SGD
ALFAL19633.03 ALFAL19633.MD
3-Bromo-5-nitrobenzoic acid ≥99%
3-Bromo-5-nitrobenzoic acid

Purity: 99% CAS No.: 6307-83-1 Molecular Formula: C7H4BrNO4 Formula Weight: 246.02 Melting Point: 159-161° Beilstein Reference: 2051365

Formula: C₇H₄BrNO₄
MW: 246,02 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00100098
CAS Number: 6307-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR