3-Bromo-2,6-dimethoxybenzoic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
A15971.03
A15971.06
ALFAA15971.03EA
0
SGD
ALFAA15971.03
ALFAA15971.06
3-Bromo-2,6-dimethoxybenzoic acid ≥98%
3-Bromo-2,6-dimethoxybenzoic acid
Purity: 98%
CAS No.: 73219-89-3
Molecular Formula: C9H9BrO4
Formula Weight: 261.07
Melting Point: 144-148°
MDL No.: MFCD00052933
Formula:
C₉H₉BrO₄ Melting Pt: 144…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052933 CAS Number: 73219-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...