2-Bromo-3,4,5,6-tetrafluorobenzoyl chloride 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Bromo-3,4,5,6-tetrafluorobenzene-1-carbonyl chloride
L20151.03
L20151.06
ALFAL20151.03EA
0
SGD
ALFAL20151.03
ALFAL20151.06
2-Bromo-3,4,5,6-tetrafluorobenzoyl chloride 97%
2-Bromo-3,4,5,6-tetrafluorobenzoyl chloride
Purity: 97%
CAS No.: 151096-42-3
Molecular Formula: C7BrClF4O
Formula Weight: 291.43
Refractive Index: 1.4980
Boiling Point: 125-126°/80mm
UN No.: 3265
MDL No.: MFCD03001174
Formula:
C₇BrClF₄O MW: 291,43 g/mol Boiling Pt: 125…126 °C (80 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD03001174 CAS Number: 151096-42-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...