3-Bromo-2,4,5-trifluorobenzoic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
L20154.03
L20154.06
ALFAL20154.03EA
0
SGD
ALFAL20154.03
ALFAL20154.06
3-Bromo-2,4,5-trifluorobenzoic acid ≥98%
3-Bromo-2,4,5-trifluorobenzoic acid
Purity: 98%
CAS No.: 104222-42-6
Molecular Formula: C7H2BrF3O2
Formula Weight: 254.99
Melting Point: 122-123°
Beilstein Reference: 7584381
Formula:
C₇H₂BrF₃O₂ MW: 254,99 g/mol Melting Pt: 122…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664065 CAS Number: 104222-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...