3-Bromophenyl isocyanate ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 1-Bromo-3-isocyanatobenzene

L12292.03 L12292.06
ALFAL12292.03EA 89 SGD
ALFAL12292.03 ALFAL12292.06
3-Bromophenyl isocyanate ≥98%
3-Bromophenyl isocyanate

Purity: 98% CAS No.: 23138-55-8 Molecular Formula: C7H4BrNO Formula Weight: 198.03 Storage Temperature: 2°C to 8°C Flash Point: 108°(226°F) Refractive Index: 1.5855 Odour: Pungent Boiling Point: 220° Beilstein Reference: 2802537 UN No.: 2206 MDL No.: MFCD00002014

Formula: C₇H₄BrNO
MW: 198,02 g/mol
Boiling Pt: 220 °C
Density: 1,586
Flash Pt: 108 °C (226 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002014
CAS Number: 23138-55-8
UN: 2206
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR