2,6-Dichlorophenylboronic acid 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(2,6-Dichlorophenyl)metaboric acid, 2,6-Dichlorobenzeneboronic acid, Dihydroxy(2,6-dichlorophenyl)borane, (2,6-Dichlorophenyl)boronic acid
L19816.03
L19816.MD
ALFAL19816.03EA
201
SGD
ALFAL19816.03
ALFAL19816.MD
2,6-Dichlorophenylboronic acid 95%
2,6-Dichlorophenylboronic acid
Purity: 95%
CAS No.: 73852-17-2
Molecular Formula: C6H5BCl2O2
Formula Weight: 190.82
Melting Point: 151-152°
Beilstein Reference: 4802506
Formula:
C₆H₅BCl₂O₂ MW: 190,82 g/mol Melting Pt: 151…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064869 CAS Number: 73852-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...