Blotting membranes, nitrocellulose
Supplier: ADVANTEC
S020A330R S045A082C S045A085C S045A154D S045A204D S045A330R
ADVTS020A330REA 507 SGD
ADVTS020A330R ADVTS045A082C ADVTS045A085C ADVTS045A154D ADVTS045A204D ADVTS045A330R
Blotting membranes, nitrocellulose
Transfer Membranes
These nitrocellulose blotting membranes are made from pure esters of nitrocellulose and are free of acetate and other esters.
  • Maximal protein/nucleic acid binding upto 80 to 100 µg/cm²
  • Low background
Available in two pore sizes: 0,20 µm (for blotting applications) and 0,45 µm (for lower molecular weights).