Glass fibre filters without binder, grade GA-55
Supplier: Advantech
GA5525MM GA5537MM GA5547MM GA5555MM GA5590MM GA55110MM GA55125MM GA55150MM GA558X10IN
ADVTGA5525MMEA 0 SGD
ADVTGA5525MM ADVTGA5537MM ADVTGA5547MM ADVTGA5555MM ADVTGA5590MM ADVTGA55110MM ADVTGA55125MM ADVTGA55150MM ADVTGA558X10IN
Glass fibre filters without binder, grade GA-55
Filters Glass Fibre Filters
These binder free glass fibre filters are widley used for air pollution monitoring and provide fast flow rates for analytical and gravimetric studies.
  • Fine porosity (0,6 micron particle retention)