5-Chloro-2-fluorobenzenesulphonyl chloride ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 5-Chloro-2-fluorobenzene-1-sulphonyl chloride

L19514.03 L19514.06
ALFAL19514.03EA 0 SGD
ALFAL19514.03 ALFAL19514.06
5-Chloro-2-fluorobenzenesulphonyl chloride ≥97%
5-Chloro-2-fluorobenzenesulphonyl chloride

Purity: 97% CAS No.: 351003-49-1 Molecular Formula: C6H3Cl2FO2S Formula Weight: 229.06 Flash Point: 110°(230°F) Melting Point: 41-44° UN No.: 3261 MDL No.: MFCD03094221

Formula: C₆H₃Cl₂FO₂S
MW: 229,06 g/mol
Melting Pt: 41…44 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03094221
CAS Number: 351003-49-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR