3,5-Dichlorophenyl isothiocyanate ≥98%
Supplier: Thermo Scientific
|
Danger
|
B20687.06
B20687.14
ALFAB20687.06EA
143
SGD
ALFAB20687.06
ALFAB20687.14
3,5-Dichlorophenyl isothiocyanate ≥98%
3,5-Dichlorophenyl isothiocyanate
Purity: 98%
CAS No.: 6590-93-8
Molecular Formula: C7H3Cl2NS
Formula Weight: 204.08
Flash Point:
Formula:
C₇H₃Cl₂NS MW: 204,08 g/mol Boiling Pt: 137…138 °C (9 mmHg) Melting Pt: 44…48 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041072 CAS Number: 6590-93-8 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...