2,6-Dichlorobenzenesulphonyl chloride ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 2,6-Dichlorobenzene-1-sulphonyl chloride

A15615.03 A15615.06
ALFAA15615.03EA 0 SGD
ALFAA15615.03 ALFAA15615.06
2,6-Dichlorobenzenesulphonyl chloride ≥97%
2,6-Dichlorobenzenesulphonyl chloride

Purity: 97% CAS No.: 6579-54-0 Molecular Formula: C6H3Cl3O2S Formula Weight: 245.51 Flash Point: 110°(230°F) Melting Point: 51-55° Beilstein Reference: 1531023 UN No.: 3261 MDL No.: MFCD00052311

Formula: C₆H₃Cl₃O₂S
MW: 245,51 g/mol
Melting Pt: 51…55 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00052311
CAS Number: 6579-54-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR